Få mere at vide
Risperidone Z-Oxime Impurity, TRC
Artikelnummer. 30134812
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30134812 25 mg -
30006019 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134812 Leverandør Toronto Research Chemicals Leverandørnr. TRCR52501625MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 132961-05-8
Molekylær formel C23 H28 F2 N4 O2
Molekylvægt (g/mol) 430.49
Synonym 3-[2-[4-[(Z)-(2,4-Difluorophenyl)(hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, Risperidone Imp. B (EP)
IUPAC navn 3-[2-[4-[(Z)-C-(2,4-difluorophenyl)-N-hydroxycarbonimidoyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMIL CC1=C(CCN2CCC(CC2)\C(=N\O)\c3ccc(F)cc3F)C(=O)N4CCCCC4=N1
Tekniske data
Kemisk navn eller materiale 3-[2-[4-[(Z)-(2,4-Difluorophenyl)(hydroxyimino)methyl]piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
Mængde 25 mg
InChI formel InChI=1S/C23H28F2N4O2/c1-15-18(23(30)29-10-3-2-4-21(29)26-15)9-13-28-11-7-16(8-12-28)22(27-31)19-6-5-17(24)14-20(19)25/h5-6,14,16,31H,2-4,7-13H2,1H3/b27-22-
Formel vægt 430.218