Få mere at vide
Risperidone EP Impurity F, Mikromol™
Artikelnummer. 30051283
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051283 25 mg
30049634 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051283 Leverandør Mikromol Leverandørnr. MM0491.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Risperidone EP Impurity F
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[2-Methyl-4-oxo-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-3-yl]ethyl 4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-carboxylate
CAS 1346603-86-8
Analyt- eller komponentnavne 2-[2-Methyl-4-oxo-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-3-yl]ethyl 4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-carboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C24 H27 F N4 O4
Certificeringer/compliance ISO 9001
Synonym Risperidone Imp. F (EP), 2-[2-Methyl-4-oxo-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-3-yl]ethyl 4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-carboxylate, Risperidone Impurity F, Risperidone Carboxylate Impurity, Risperidone EP Impurity F
InChI formel InChI=1S/C24H27FN4O4/c1-15-18(23(30)29-10-3-2-4-21(29)26-15)9-13-32-24(31)28-11-7-16(8-12-28)22-19-6-5-17(25)14-20(19)33-27-22/h5-6,14,16H,2-4,7-13H2,1H3
SMIL CC1=C(CCOC(N2CCC(C3=NOC4=C3C=CC(F)=C4)CC2)=O)C(N5CCCCC5=N1)=O
IUPAC navn 2-(2-methyl-4-oxo-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-3-yl)ethyl 4-(6-fluorobenzo[d]isoxazol-3-yl)piperidine-1-carboxylate
Molekylvægt (g/mol) 454.5
Mængde 25 mg
Formel vægt 454.2016 g/mol