Få mere at vide
Risperidone Carboxylate Impurity, TRC
Artikelnummer. 30179984
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179984 100 mg
30179769 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179984 Leverandør Toronto Research Chemicals Leverandørnr. TRCR525025100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346603-86-8
Molekylær formel C24 H27 F N4 O4
Molekylvægt (g/mol) 454.5
Synonym 2-[2-Methyl-4-oxo-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-3-yl]ethyl 4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-carboxylate, Risperidone Imp. F (EP), 2-(6,7,8,9-Tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-3-yl)ethyl 4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinecarboxylate (ACI), 1-Piperidinecarboxylic acid, 4-(6-fluoro-1,2-benzisoxazol-3-yl)-, 2-(6,7,8,9-tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-3-yl)ethyl ester (ACI)
IUPAC navn 2-(2-methyl-4-oxo-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-3-yl)ethyl 4-(6-fluoro-1,2-benzoxazol-3-yl)piperidine-1-carboxylate
SMIL CC1=C(CCOC(=O)N2CCC(CC2)c3noc4cc(F)ccc34)C(=O)N5CCCCC5=N1
Tekniske data
Kemisk navn eller materiale 2-[2-Methyl-4-oxo-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-3-yl]ethyl 4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-carboxylate
Mængde 100 mg
InChI formel InChI=1S/C24H27FN4O4/c1-15-18(23(30)29-10-3-2-4-21(29)26-15)9-13-32-24(31)28-11-7-16(8-12-28)22-19-6-5-17(25)14-20(19)33-27-22/h5-6,14,16H,2-4,7-13H2,1H3
Formel vægt 454.2016