Få mere at vide
Rislenemdaz, MedChemExpress
Artikelnummer. 16496787
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16496787 5 mg
16464088 10 mg
16461069 1 mg
16440447 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16496787 Leverandør Medchem Express Leverandørnr. HY106441A
MedChemExpress Rislenemdaz (CERC-301) is an orally bioavailable and selective N-methyl-D-aspartate (NMDA) receptor subunit 2B (GluN2B) antagonist with Ki and IC 50 of 8.1 nM and 3.6 nM, respectively.
Rislenemdaz (CERC-301) is an orally bioavailable and selective N-methyl-D-aspartate (NMDA) receptor subunit 2B (GluN2B) antagonist with Ki and IC 50 of 8.1 nM and 3.6 nM, respectively.
Tekniske data
Kemisk navn eller materiale Rislenemdaz
Fysisk form Solid
CAS 808732-98-1
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C19H23FN4O2
Til brug med (applikation) Neuroscience-Neurodegeneration
Synonym MK-0657 CERC-301
Opløselighedsinformation DMSO : 100 mg/mL (279.01 mM; Need ultrasonic)
SMIL O=C(N1C[C@@H](F)[C@@H](CNC2=NC=CC=N2)CC1)OCC3=CC=C(C)C=C3
Molekylvægt (g/mol) 358.41
Formel vægt 358.41
Procent renhed 97.5%