Få mere at vide
Risedronate Sodium 2.5 Hydrate, LoGiCal
Artikelnummer. 30457568
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30457568 10 mg
1 options
Artikelnummer. 30457568 Leverandør LoGiCal Leverandørnr. LGCFOR0777.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Risedronate Sodium 2.5 Hydrate
CAS 329003-65-8
Analyt- eller komponentnavne Risedronate Sodium 2.5 Hydrate
Forsendelsestilstand Room Temperature
Molekylær formel 2 C7 H10 N O7 P2 . 2 Na . 5 H2 O
Certificeringer/compliance ISO 9001
Synonym Risedronate Sodium 2.5 Hydrate, P,P'-[1-Hydroxy-2-(3-pyridinyl)ethylidene]bisphosphonic acid sodium salt hydrate (2:2:5), [1-Hydroxy-2-(3-pyridinyl)ethylidene]bisphosphonic acid monosodium salt hydrate (2:5), Risedronic acid monosodium salt hemipentahydrate
InChI formel InChI=1S/2C7H11NO7P2.2Na.5H2O/c2*9-7(16(10,11)12,17(13,14)15)4-6-2-1-3-8-5-6;;;;;;;/h2*1-3,5,9H,4H2,(H2,10,11,12)(H2,13,14,15);;;5*1H2/q;;2*+1;;;;;/p-2
Løsningstype Neat
SMIL O.O.O.O.O.[Na+].[Na+].OC(Cc1cccnc1)(P(=O)(O)O)P(=O)(O)[O-].OC(Cc2cccnc2)(P(=O)(O)O)P(=O)(O)[O-]
IUPAC navn disodium;hydroxy-(1-hydroxy-1-phosphono-2-pyridin-3-ylethyl)phosphinate;pentahydrate
Industry Type Clinical Testing
Molekylvægt (g/mol) 700.26
Mængde 10 mg
Formel vægt 700.0189 g/mol