Learn More
Invitrogen™ Human RIPK3, GST Tag Recombinant Protein
RIPK3 is a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases. Recombinant full-length human RIPK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Brand: Invitrogen™ PV6397
Additional Details : Weight : 0.10000kg
Description
RIPK3 contains a C-terminal domain unique from other RIP family members. RIPK3 is predominantly localized to the cytoplasm, and specifically, are colocalized in the mitochondrion which can undergo nucleocytoplasmic shuttling dependent on novel nuclear localization and export signals. RIPK3 is a component of the tumor necrosis factor (TNF) receptor-I signaling complex, and can induce apoptosis and weakly activate the NF-kappaB transcription factor and caspase-8 prevents RIPK3-dependent necrosis without inducing apoptosis by functioning in a proteolytically active complex with CFLAR and that this complex is required for the protective function.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
83.2 kDa | |
Human | |
GST Tag | |
Biochemical binding/kinetic | |
RIPK3 | |
Kinases & Inhibitors | |
Liquid |
10μg | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
|
Serine/Threonine Kinase | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |