Learn More
Emma Safety Footwear Ringo XD Safety Shoes
Product Code. 15998951
Change view
Click to view available options
Size:
44
49
Unit Size:
1 pair
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998951 49 1 pair
13266564 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15998951 Supplier Emma Safety Footwear Supplier No. MM73156049
  • ONLINE EXCLUSIVE PRICE
    970.64 DKK
    572.25 DKK / 1 par
    Spare 398.39 DKK
    41% Slukket
Forventet forsendelse: 16-03-2027
A versatile shoe with wipe clean uppers and an 'easy clean' sole. Popular with people working in Food and Catering. For peace of mind, an internal toe cap provides resilient protection against falling objects and unforeseen hazards.
  • S2-Safety shoe for wet working conditions
  • Full grain leather construction
  • GripForce™ Easy Twist Outsole
  • Hydro-Tec™ Sanitized Silver
Specifications
Design Ringo XD
Material Leather
Size 49