Få mere at vide
Rimantadine-d4 Hydrochloride, TRC
Artikelnummer. 30418699
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30418699 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418699 Leverandør Toronto Research Chemicals Leverandørnr. TRCR5170025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rimantadine-d4 Hydrochloride
CAS 350818-67-6
Renhedsgrad noter HPLC
Molekylær formel C12 D4 H17 N . Cl H
Mængde 5 mg
Synonym Tricyclo[3.3.1.13,7]decane-1-methan-α-d-amine','α-(methyl-d3)-','hydrochloride (9CI), α-(Methyl-d3)tricyclo[3.3.1.13,7]decane-1-methan-α-d-amine hydrochloride, (±)-Rimantadine-d4 hydrochloride, 1-(1-Adamantyl)ethylamine-1,2,2,2-d4 hydrochloride, 1-(1-Aminoethyl-1,2,2,2-d4)adamantane hydrochloride, 1-(Adamantan-1-yl)ethanamine-d4 hydrochloride, 1-Rimantadine-d4 hydrochloride, Rimantadine-d4 hydrochloride, Rimantadine-D4 (aminoethyl-1,2,2,2-D4) hydrochloride
InChI formel InChI=1S/C12H21N.ClH/c1-8(13)12-5-9-2-10(6-12)4-11(3-9)7-12;/h8-11H,2-7,13H2,1H3;1H/i1D3,8D;
SMIL Cl.[2H]C([2H])([2H])C([2H])(N)C12CC3CC(CC(C3)C1)C2
IUPAC navn 1-(1-adamantyl)-1,2,2,2-tetradeuterioethanamine;hydrochloride
Molekylvægt (g/mol) 219.787
Formel vægt 219.169
Procent renhed >95