Få mere at vide
Rilpivirine Hydrochloride, Mikromol™
Artikelnummer. 17992676
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
17992676 100 mg
1 options
Artikelnummer. 17992676 Leverandør Mikromol Leverandørnr. MM3376.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Rilpivirine Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rilpivirine Hydrochloride
CAS 700361-47-3
Analyt- eller komponentnavne Rilpivirine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C22 H18 N6 . Cl H
Certificeringer/compliance ISO 17034
Synonym Rilpivirine Hydrochloride, Benzonitrile, 4-[[4-[[4-[(1E)-2-cyanoethenyl]-2,6-dimethylphenyl]amino]-2-pyrimidinyl]amino]-, hydrochloride (1:1), Benzonitrile, 4-[[4-[[4-[(1E)-2-cyanoethenyl]-2,6-dimethylphenyl]amino]-2-pyrimidinyl]amino]-, monohydrochloride (9CI), Rilpirivine, Rilpivirine hydrochloride, TMC 278 hydrochloride, R314585
InChI formel InChI=1S/C22H18N6.ClH/c1-15-12-18(4-3-10-23)13-16(2)21(15)27-20-9-11-25-22(28-20)26-19-7-5-17(14-24)6-8-19;/h3-9,11-13H,1-2H3,(H2,25,26,27,28);1H/b4-3+;
SMIL Cl.Cc1cc(\C=C\C#N)cc(C)c1Nc2ccnc(Nc3ccc(cc3)C#N)n2
IUPAC navn 4-[[4-[4-[(E)-2-cyanoethenyl]-2,6-dimethylanilino]pyrimidin-2-yl]amino]benzonitrile;hydrochloride
Molekylvægt (g/mol) 402.88
Mængde 100 mg
Formel vægt 402.136 g/mol