missing translation for 'onlineSavingsMsg'
Learn More

Rilmenidine Hemifumarate, TRC

Product Code. 17932305
Change view
Click to view available options
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17932305 1 mg
17911635 5 mg
17990455 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17932305 Supplier Toronto Research Chemicals Supplier No. TRCR5097051MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207572-68-7
Molekylær formel 2 C10 H16 N2 O . C4 H4 O4
Molekylvægt (g/mol) 476.566
Synonym N-(Dicyclopropylmethyl)-4,5-dihydro-2-oxazolamine (E)-2-Butenedioate, S 3341, Oxaminozoline;
IUPAC navn (E)-but-2-enedioic acid;N-(dicyclopropylmethyl)-4,5-dihydro-1,3-oxazol-2-amine
SMIL OC(=O)\C=C\C(=O)O.C1CN=C(NC(C2CC2)C3CC3)O1.C4CN=C(NC(C5CC5)C6CC6)O4

Specifications

Kemisk navn eller materiale Rilmenidine Hemifumarate
Mængde 1 mg
InChI formel InChI=1S/2C10H16N2O.C4H4O4/c2*1-2-7(1)9(8-3-4-8)12-10-11-5-6-13-10;5-3(6)1-2-4(7)8/h2*7-9H,1-6H2,(H,11,12);1-2H,(H,5,6)(H,7,8)/b;;2-1+
Formel vægt 476.263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.