Få mere at vide
Tocris Bioscience™ Rilmenidine hemifumarate
Beskrivelse
Rilmenidine hemifumarate is an I1-imidazoline binding site selective ligand and α2-adrenoceptor agonist. Possesses greater I1 vs α2 selectivity than the prototypical compound, clonidine. Also thought to enhance autophagy; shown to increase LC3-II levels in PC12 cells.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-[N-(Dicyclopropylmethyl)amino]oxazoline hemifumarate |
| CAS | 207572-68-7 |
| Analyseprocentområde | >99% |
| Mængde | 5 mg |
| Molekylær formel | C10H16N2O.½C4H4O4 |
| Formel vægt | Observed MW: 238.29 |
| Renhed | 0.98 |