Få mere at vide
Rilmenidine Dihydrogen Phosphate, Mikromol™
Beskrivelse
Rilmenidine Dihydrogen Phosphate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Rilmenidine Dihydrogen Phosphate |
| CAS | 85409-38-7 |
| Analyt- eller komponentnavne | Rilmenidine Dihydrogen Phosphate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C10 H16 N2 O . H3 O4 P |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Rilmenidine dihydrogen phosphate, N-(Dicyclopropylmethyl)-4,5-dihydro-oxazol-2-amine dihydrogen phosphate (1:1), S 3341-3, Rilmenidine for system suitability |
| InChI formel | InChI=1S/C10H16N2O.H3O4P/c1-2-7(1)9(8-3-4-8)12-10-11-5-6-13-10;1-5(2,3)4/h7-9H,1-6H2,(H,11,12);(H3,1,2,3,4) |
| SMIL | OP(=O)(O)O.C1CN=C(NC(C2CC2)C3CC3)O1 |
| IUPAC navn | N-(dicyclopropylmethyl)-4,5-dihydro-1,3-oxazol-2-amine;phosphoric acid |
| Vis mere |