Få mere at vide
Rilmazafone Hydrochloride, TRC
Artikelnummer. 30008899
Skift visning
Mængde:
0.5 mg
2.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30008899 2.5 mg 2.5mg
30006558 5 mg -
30005288 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008899 Leverandør Toronto Research Chemicals Leverandørnr. TRCR5097152.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85815-37-8
Molekylær formel C21H20Cl2N6O3 . HCl
Molekylvægt (g/mol) 511.8
Synonym 5-[[(Aminoacetyl)amino]methyl]-1-[4-chloro-2-(2-chlorobenzoyl)phenyl]-N,N-dimethyl-1H-1,2,4-triazole-3-carboxamide Monohydrochloride, 450191S
IUPAC navn 5-[[(2-aminoacetyl)amino]methyl]-1-[4-chloro-2-(2-chlorobenzoyl)phenyl]-N,N-dimethyl-1,2,4-triazole-3-carboxamide;hydrochloride
SMIL Cl.CN(C)C(=O)c1nc(CNC(=O)CN)n(n1)c2ccc(Cl)cc2C(=O)c3ccccc3Cl
Tekniske data
Kemisk navn eller materiale Rilmazafone Hydrochloride
Mængde 2.5 mg
InChI formel InChI=1S/C21H20Cl2N6O3.ClH/c1-28(2)21(32)20-26-17(11-25-18(30)10-24)29(27-20)16-8-7-12(22)9-14(16)19(31)13-5-3-4-6-15(13)23;/h3-9H,10-11,24H2,1-2H3,(H,25,30);1H
Formel vægt 510.0741