missing translation for 'onlineSavingsMsg'
Learn More
Rilematovir, MedChemExpress Product Code.: 16483146

Rilematovir, MedChemExpress

Product Code. p-200099653
25 mg, 25mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483146

Brand: Medchem Express HY112180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rilematovir (JNJ-678) is a novel fusion protein inhibitor. Rilematovir has the potential for respiratory syncytial virus (RSV) research.

Rilematovir (JNJ-678) is a novel fusion protein inhibitor. Rilematovir has the potential for respiratory syncytial virus (RSV) research.

Specifications

Chemical Name or Material Rilematovir
CAS 1383450-81-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H20ClF3N4O3S
Quantity 25 mg
Synonym JNJ-678 JNJ-53718678
Solubility Information DMSO : 65 mg/mL (129.76 mM; Need ultrasonic)
SMILES O=C(N1CC(F)(F)F)N(CC(N2CCCS(=O)(C)=O)=CC3=C2C=CC(Cl)=C3)C4=C1C=CN=C4
Molecular Weight (g/mol) 500.92
Formula Weight 500.92
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Rilematovir, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.