Få mere at vide
Rifampicin-D3, TRC
Artikelnummer. 30413631
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30413631 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413631 Leverandør Toronto Research Chemicals Leverandørnr. TRCR5080020.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rifampicin-d3
CAS 1262052-36-7
Renhedsgrad noter HPLC
Molekylær formel C43H55D3N4O12
Mængde 0.5 mg
Synonym 3-[[(4-Methyl-d3-1-piperazin-yl)imino]methyl]rifamycin, Rifampin-d3, Rifaldazine-d3, Rifamycin AMP-d3, R/AMP-d3, Abrifam, -d3 Eremfat-d3, Rifa-d3
InChI formel InChI=1S/C43H58N4O12/c1-21-12-11-13-22(2)42(55)45-33-28(20-44-47-17-15-46(9)16-18-47)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,59-40)57-19-14-29(56-10)23(3)39(58-27(7)48)25(5)35(50)24(4)34(21)49/h11-14,19-21,23-25,29,34-35,39,49-53H,15-18H2,1-10H3,(H,45,55)/b12-11+,19-14+,22-13-,44-20+/t21-,23+,24+,25+,29-,34-,35+,39+,43-/m0/s1/i9D3
SMIL OC1=C(/C=N/N2CCN(C([2H])([2H])[2H])CC2)C3=C(O)C4=C(O)C(C)=C(O[C@@](O/C=C/[C@H](OC)[C@@H](C)[C@@H](OC(C)=O)[C@H](C)[C@H](O)[C@H](C)[C@@H](O)[C@@H](C)/C=C/C=C(C)/C(N3)=O)(C)C5=O)C5=C41
IUPAC navn [(7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-26-[(E)-[4-(trideuteriomethyl)piperazin-1-yl]iminomethyl]-8,30-dioxa-24-azatetracyclo[23.3.1.14,7.05,28]triaconta-1(28),2,4,9,19,21,25(29),26-octaen-13-yl] acetate
Molekylvægt (g/mol) 825.96
Formel vægt 825.424
Procent renhed >95