Få mere at vide
Ricolinostat, MedChemExpress
Artikelnummer. 16486968
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16486968 10 mg -
16453027 50 mg -
16439517 5 mg -
16447046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16486968 Leverandør Medchem Express Leverandørnr. HY16026
MedChemExpress Ricolinostat (ACY-1215) is a potent and selective HDAC6 inhibitor, with an IC50 of 5 nM. ACY-1215 also inhibits HDAC1, HDAC2, and HDAC3 with IC50s of 58, 48, and 51 nM, respectively.
Ricolinostat (ACY-1215) is a potent and selective HDAC6 inhibitor, with an IC50 of 5 nM. ACY-1215 also inhibits HDAC1, HDAC2, and HDAC3 with IC50s of 58, 48, and 51 nM, respectively.
Tekniske data
Kemisk navn eller materiale Ricolinostat
CAS 1316214-52-4
Farve White
Renhedsgrad noter Research
Molekylær formel C24H27N5O3
Mængde 10 mg
Synonym ACY-1215 Rocilinostat
Opløselighedsinformation DMSO : ≥ 50 mg/mL (115.34 mM)
SMIL O=C(C1=CN=C(N(C2=CC=CC=C2)C3=CC=CC=C3)N=C1)NCCCCCCC(NO)=O
Molekylvægt (g/mol) 433.5
Formel vægt 433.5
Procent renhed 95.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death