Få mere at vide
Ricinolic Acid (~80%), TRC
Artikelnummer. 30134111
Skift visning
Mængde:
100 mg
500 mg
1 g
Pakningsstørrelse:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30134111 1 g -
17991925 500 mg 0.5mg
17974615 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134111 Leverandør Toronto Research Chemicals Leverandørnr. TRCR4956001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 141-22-0
Molekylær formel C18 H34 O3
Molekylvægt (g/mol) 298.46
Synonym 9-Octadecenoic acid, 12-hydroxy-, (9Z,12R)-, 9-Octadecenoic acid, 12-hydroxy-, [R-(Z)]-, Ricinoleic acid (8CI), (9Z,12R)-12-Hydroxy-9-octadecenoic acid, (9Z,12R)-12-Hydroxyoctadec-9-enoic acid, (R)-12-Hydroxyoleic acid, (R)-Ricinoleic acid, 12-D-Hydroxy-9-cis-octadecenoic acid, 12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoctadeca-9-enoic acid, CS 80, Cenwax C, Edenor Ri 90, H 1657, NSC 281242, Nouracid CS 80, Ricinic acid, Ricinolic acid
IUPAC navn (Z,12R)-12-hydroxyoctadec-9-enoic acid
SMIL CCCCCC[C@@H](O)C\C=C/CCCCCCCC(=O)O
Tekniske data
Kemisk navn eller materiale Ricinoleic acid
Mængde 1 g
InChI formel InChI=1S/C18H34O3/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21/h9,12,17,19H,2-8,10-11,13-16H2,1H3,(H,20,21)/b12-9-/t17-/m1/s1
Formel vægt 298.2508