Få mere at vide
Ricinine-d3, TRC
Artikelnummer. 30419925
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30419925 2 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419925 Leverandør Toronto Research Chemicals Leverandørnr. TRCR4955522MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ricinine-d3
CAS 1313734-77-8
Molekylær formel C8 H5 D3 N2 O2
Mængde 2 mg
Synonym 1,2-Dihydro-4-methoxy-1-(methyl-d3)-2-oxo-3-pyridinecarbonitrile, 1,2-Dihydro-4-methoxy-1-(methyl-d3)2-oxonicotinonitrile, Recinine-d3, NSC 409913-d3, NSC 642604-d3, Ricinin-d3
InChI formel InChI=1S/C8H8N2O2/c1-10-4-3-7(12-2)6(5-9)8(10)11/h3-4H,1-2H3/i1D3
SMIL [2H]C([2H])([2H])N1C=CC(=C(C#N)C1=O)OC
IUPAC navn 4-methoxy-2-oxo-1-(trideuteriomethyl)pyridine-3-carbonitrile
Molekylvægt (g/mol) 167.1806
Formel vægt 167.0774