Learn More
Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver, Silver Nitrate #2 Solution
Product Code. 12543657 Shop All Epredia Products
Silver Nitrate Solution #1, 500mL
Click to view available options
Type:
Gum Mastic Solution
Hydroquinone
Sensitizer Solution
Silver Nitrate Solution #1
Steiner-Steiner Silver Kit
Unit Size:
500mL
Each
Pack of 12
This item is not returnable. View return policy
This item is not returnable. View return policy
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver.
  • Kit does not contain uranyl nitrate
  • Background is stained yellow to brown
  • Bacteria, Donovan bodies and fungi all stain black
  • Kit formulation and detailed procedures provide consistent and reproducible results
TRUSTED_SUSTAINABILITY
Specifications
Type Silver Nitrate Solution #1
Quantity 500mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver, Silver Nitrate #2 Solution >