Learn More
Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver, Silver Nitrate #2 Solution Product Code.: 12543657
Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver, Silver Nitrate #2 Solution
Product Code. 12543657 Shop All Epredia Products
Silver Nitrate Solution #1, 500mL
Click to view available options
Type:
Gum Mastic Solution
Hydroquinone
Sensitizer Solution
Silver Nitrate Solution #1
Steiner-Steiner Silver Kit
Unit Size:
500mL
Each
Pack of 12
Product Code. 12543657
Brand: Epredia™ 88033
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver.
  • Kit does not contain uranyl nitrate
  • Background is stained yellow to brown
  • Bacteria, Donovan bodies and fungi all stain black
  • Kit formulation and detailed procedures provide consistent and reproducible results
TRUSTED_SUSTAINABILITY
Specifications
Type Silver Nitrate Solution #1
Quantity 500mL
Product Content Correction
Product Title
Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver, Silver Nitrate #2 Solution >