Learn More
Epredia™ STS™ Specimen Transport System Pre-filled Specimen Containers 10% NBF - Bio-Tite™
Transport, store and retrieve pathology specimens in a safe and cost effective manner with Epredia™ Specimen Transportation System (STS™).
POA
Specifications
Product Type | Pre-filled Specimen Container |
---|---|
Material | Polypropylene |
Sterility | Non-sterile |
Closure Type | Bio-Tite Lid |
Color | Clear |
Specifications
Pre-filled Specimen Container | |
Non-sterile | |
Clear |
Polypropylene | |
Bio-Tite Lid | |
10% NBF |