Få mere at vide
Ribavirin EP Impurity D, Mikromol™
Artikelnummer. 30461043
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30461043 4 x 25 mg
30050904 25 mg
17983466 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30461043 Leverandør Mikromol Leverandørnr. MM0542.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ribavirin EP Impurity D, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1H-1,2,4-Triazole-3-carboxamide
CAS 8-5-3641
Analyt- eller komponentnavne 1H-1,2,4-Triazole-3-carboxamide
Forsendelsestilstand Room Temperature
Molekylær formel C3 H4 N4 O
Certificeringer/compliance ISO 9001
Synonym Ribavirin Imp. D (EP), Ribavirin USP Related Compound D, Ribavirin USP RC D, 1H-1,2,4-Triazole-3-carboxamide, Ribavirin Related Compound D, Ribavirin Impurity D
InChI formel InChI=1S/C3H4N4O/c4-2(8)3-5-1-6-7-3/h1H,(H2,4,8)(H,5,6,7)
Løsningstype Neat
SMIL NC(=O)c1nc[nH]n1
IUPAC navn 1H-1,2,4-triazole-3-carboxamide
Molekylvægt (g/mol) 112.09
Mængde 4 x 25 mg
Formel vægt 112.0385 g/mol