Learn More
Rhodium(III) Chloride Hydrate, TRC
Product Code. 30408901
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30408901 1 g 1g
1 options
Product Code. 30408901 Supplier Toronto Research Chemicals Supplier No. TRCR3186451G
 Restricted Item
Forventet forsendelse: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Rhodium(III) Chloride Hydrate
CAS 20765-98-4
Molecular Formula Cl3Rh.xH2O
Quantity 1 g
Synonym Rhodium Trichloride Hydrate
InChI Formula InChI=1S/C21H27F3O8S/c1-5-6-7-10-15(25)11-8-9-14(2)31-20(26)19-17(30-4)12-16(29-3)13-18(19)32-33(27,28)21(22,23)24/h5,12-14H,1,6-11H2,2-4H3
SMILES COc1cc(OC)c(C(=O)OC(C)CCCC(=O)CCCC=C)c(OS(=O)(=O)C(F)(F)F)c1
IUPAC Name 6-oxoundec-10-en-2-yl 2,4-dimethoxy-6-(trifluoromethylsulfonyloxy)benzoate
Molecular Weight (g/mol) 209.26
Formula Weight 496.1379