missing translation for 'onlineSavingsMsg'
Få mere at vide

RhoA Antibody (1B3-4A10), Novus Biologicals™

Artikelnummer. 18713404 Shop alle Bio Techne produkter
Change view
Click to view available options
Mængde:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18713404 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18713404 Supplier Novus Biologicals Supplier No. NBP2225290.025mg

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RhoA Monoclonal specifically detects RhoA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoA
Applikationer Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon 1B3-4A10
Koncentration 1 mg/mL
Konjugeret Unconjugated
Fortynding Western Blot 1:500 - 1:1500, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:50 - 1:500
Gene Alias h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA
Gensymboler RHOA
Værtsarter Mouse
Immunogen Full-length human recombinant RhoA protein.
Oprensningsmetode Protein A purified
Mængde 0.025 mg
Regulatorisk status RUO
Forskningsdisciplin Cancer, mTOR Pathway, Signal Transduction, Wnt Signaling Pathway
Primær eller sekundær Primary
Gen-id (Entrez) 387
Målarter Human, Mouse
Indhold og opbevaring Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.