Få mere at vide
R&D Systems™ Rhesus Macaque CD58/LFA-3 (NP_001254714) VersaClone cDNA
Artikelnummer. 18715978
Skift visning
Mængde:
10 μg
Artikelnummer. Mængde
18715978 10 μg
1 options
Artikelnummer. 18715978 Leverandør R&D Systems™ Leverandørnr. RDC2419
  • ONLINE EXCLUSIVE PRICE
    2605.00 DKK
    2460.15 DKK / 10µg
    Spare 144.85 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 28-08-2026
Læg i indkøbskurv
Læg i indkøbskurv
R&D Systems™ Rhesus Macaque CD58/LFA-3 (NP_001254714) VersaClone cDNA is a member of the CD2 family of the immunoglobulin superfamily.
The Rhesus Macaque CD58/LFA-3 (NP_001254714) VersaClone cDNA clone contains the complete ORF for CD58/LFA-3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD58/LFA-3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY
Tekniske data
Gene rhesusCD58
Adgangsnummer NP_001254714
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl
Indsæt type 766bp
pH 8.5
Gen-id (Entrez) 965
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Rhesus Macaque
Mængde 10 μg