missing translation for 'onlineSavingsMsg'
Få mere at vide

Rheinanthrone, TRC

Artikelnummer. 30413873
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30413873 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413873 Supplier Toronto Research Chemicals Supplier No. TRCR31501050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Rheinanthrone
CAS 480-09-1
Renhedsgrad noter HPLC
Molekylær formel C15 H10 O5
Mængde 50 mg
Synonym 9,10-dihydro-4,5-dihydroxy-10-oxo-2-anthroic Acid, 1,8-Dihydroxy-9-anthrone-3-carboxylic acid, NSC 658580, Rhein-9-anthrone
InChI formel InChI=1S/C15H10O5/c16-10-3-1-2-7-4-8-5-9(15(19)20)6-11(17)13(8)14(18)12(7)10/h1-3,5-6,16-17H,4H2,(H,19,20)
SMIL OC(=O)c1cc(O)c2C(=O)c3c(O)cccc3Cc2c1
IUPAC navn 4,5-dihydroxy-10-oxo-9H-anthracene-2-carboxylic acid
Molekylvægt (g/mol) 270.237
Formel vægt 270.053
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.