Learn More
Thermo Scientific™ RheBuild™ Kits
Product Code. 11850561
Change view
Click to view available options
For Use With (Equipment):
Rheodyne Injectors 3725/3725i/3725-038/3725i-038
Rheodyne Injectors 7010/7000
Rheodyne Injectors 7125/7126
Rheodyne Injectors 7725/7725i/7726
Rheodyne Injectors 8125/8126
Rheodyne Injectors 9125/9126
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11850561 Rheodyne Injectors 7725/7725i/7726 Each
10149464 Rheodyne Injectors 3725/3725i/3725-038/3725i-038 Each
10512645 Rheodyne Injectors 7010/7000 Each
10416684 Rheodyne Injectors 7125/7126 Each
10754956 Rheodyne Injectors 8125/8126 Each
10735126 Rheodyne Injectors 9125/9126 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11850561 Supplier Thermo Scientific™ Supplier No. 7725999
Forventet forsendelse: 26-11-2026
Everything you need to maintain your Thermo Scientific™ Rheodyne™ valves and injectors in your HPLC system.
Use Thermo Scientific™ RheBuild Kits to maintain your Thermo Scientific™ Rheodyne™ valves and injectors for high-performance liquid chromatography (HPLC). A single kit includes everything you need.
TRUSTED_SUSTAINABILITY
Specifications
Type Maintenance Kit
Description For Rheodyne models 7725/7725 i/7726
For Use With (Equipment) Rheodyne Injectors 7725/7725i/7726