missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGL4 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-05804C
This item is not returnable.
View return policy
Description
RGL4 Polyclonal specifically detects RGL4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
RGL4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
MGC119678, ral guanine nucleotide dissociation stimulator-like 4, RalGDS related oncogene, Ral-GDS related protein Rgr, ralGDS-like 4, ral-GDS-related protein, Rgr | |
Produced in rabbits immunized with E. coli-derived Human RGL4 fragment. | |
0.1 mL | |
Signal Transduction | |
266747 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction