missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
RG7800, MedChemExpress
Beskrivelse
RG7800 is a SMN2 splicing modifier. RG7800 has the potential for spinal muscular atrophy treatment.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | RG7800 |
| Fysisk form | Solid |
| CAS | 1449598-06-4 |
| Farve | Yellow |
| Mængde | 5 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C24H28N6O |
| Synonym | RO6885247 |
| Opløselighedsinformation | Ethanol : 2.5 mg/mL (6.00 mM; Need ultrasonic) ∣DMSO : 1.4 mg/mL (3.36 mM; Need ultrasonic and warming) ∣H2O : 1.25 mg/mL (3.00 mM; ultrasonic and adjust pH to 3 with HCl) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMIL | O=C1C=C(C2=NN3C(C(CC)=NC(C)=C3)=C2)N=C4N1C=C(C5CCN(C)CC5)C=C4C |
| Vis mere |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?