Learn More
Hamilton RFID Reader
Product Code. 31257794
Change view
Click to view available options
For Use With (Equipment):
Microlab 700 Instrument
Product Code. For Use With (Equipment)
31257794 Microlab 700 Instrument
1 options
Product Code. 31257794 Supplier Hamilton Supplier No. 10106736
 Restricted Item
Forventet forsendelse: 03-12-2026
RFID reader designed for use with compatible laboratory systems. Enables identification and tracking functionality for improved workflow.
  • Microlab RFID Reader (PN: 10106736) for secure user authentication
  • Enables controlled access to the Microlab 700 system
  • Connects to the Microlab 700 controller via USB
  • Provides fast and reliable user verification using RFID technology
Specifications
Product Type RFID Reader
For Use With (Equipment) Microlab 700 Instrument