Learn More
RETSCH Test Sieves - 14 Mesh or Greater
Stainless steel with wire gauze
Brand: RETSCH 60.150.000212
This item is not returnable.
View return policy
Specifications
2 in. | |
212 μm | |
ASTM E11 | |
Stainless Steel | |
No. 70 | |
8 in. |
5.08 cm | |
Test Sieve | |
8 in. | |
70 Mesh | |
Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc. | |
Stainless Steel |
Product Content Correction
Product Title
RETSCH Test Sieves - 14 Mesh or Greater > Pore Size: 212 μm, 70 Mesh
Spot an opportunity for improvement?Share a Content Correction