Learn More
RETSCH Accessories for ZM200 Ultra Centrifugal Mill Cyclone Accessory
For use with ZM200 Ultra Centrifugal Mill
Brand:  RETSCH 22.935.0016
Additional Details : Weight : 1.96000kg
Product Code. 11974272
  • 22870.00 DKK / Each
Estimated Shipment: 22-07-2024
to see stock.
Add to basket
Description
Description
  • Includes cassette pan with outlet and support
  • With filter bag
  • 5L collecting receptacle
Specifications
Specifications
Cyclone
5 L
ZM200 ultra centrifugal mil