Få mere at vide
Retigabine-d4 Dihydrochloride, TRC
Artikelnummer. 30419382
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30419382 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419382 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1890522.5MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Retigabine-d4 Dihydrochloride
CAS 1285886-70-5
Molekylær formel C16H16D4Cl2FN3O2
Mængde 2.5 mg
Synonym N-[2-Amino-4-[4-(fluorobenzyl-d4)amino]phenyl]carbamic Acid Ethyl Ester Dihydrochloride, Ethyl [2-Amino-4-[[(4-fluorophenyl-d4)methyl]amino]phenyl] carbamate Dihydrochloride, D 20443-d4
InChI formel InChI=1S/C16H18FN3O2/c1-2-22-16(21)20-15-8-7-13(9-14(15)18)19-10-11-3-5-12(17)6-4-11/h3-9,19H,2,10,18H2,1H3,(H,20,21)
SMIL FC1=C([2H])C([2H])=C(CNC2=CC=C(NC(OCC)=O)C(N)=C2)C([2H])=C1[2H].Cl.Cl
Molekylvægt (g/mol) 380.28
Formel vægt 379.1168