Få mere at vide
BD Biosciences™ Reticulocyte Reagent System (CE/IVD)
Artikelnummer. 15896148
Mængde:
200 tests
Artikelnummer. 15896148
Brand: BD Biosciences™ 349204
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
For determining reticulocytes as a percentage of total erythrocytes in human peripheral blood.
  • BD Retic-Count™ reagent is intended for determining reticulocytes as a percentage of total erythrocytes in human peripheral blood by a BD FACSCalibur™, BD FACSort™, BD FACScan™, or BD FACStrak™ flow cytometer.
  • The BD Retic-Count reagent is for in vitro diagnostic use.
TRUSTED_SUSTAINABILITY
Tekniske data
Indhold og opbevaring The Retic-COUNT reagent is light sensitive and must be stored in the dark at 2°C to 8°C. Avoid unnecessary exposure of the reagent or reagent/cell mixture to light. The reagent bottle must be kept well sealed when not in use.
Antal prøver 200 Tests
Mængde 200 tests
Stabilitet When stored at 2°C to 8°C, the reagent is stable until the expiration date shown on the label. Do not use after the expiration date.