Få mere at vide
BD Biosciences™ Reticulocyte Reagent System (CE/IVD)
Mængde:
200 tests
Beskrivelse
- BD Retic-Count™ reagent is intended for determining reticulocytes as a percentage of total erythrocytes in human peripheral blood by a BD FACSCalibur™, BD FACSort™, BD FACScan™, or BD FACStrak™ flow cytometer.
- The BD Retic-Count reagent is for in vitro diagnostic use.
Tekniske data
Tekniske data
| Indhold og opbevaring | The Retic-COUNT reagent is light sensitive and must be stored in the dark at 2°C to 8°C. Avoid unnecessary exposure of the reagent or reagent/cell mixture to light. The reagent bottle must be kept well sealed when not in use. |
| Antal prøver | 200 Tests |
| Mængde | 200 tests |
| Stabilitet | When stored at 2°C to 8°C, the reagent is stable until the expiration date shown on the label. Do not use after the expiration date. |