Få mere at vide
Reticuline Hydrochloride Salt (>90% ee), TRC
Artikelnummer. 30134749
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30134749 5 mg -
30005160 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134749 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1885005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 903-91-3
Molekylær formel C19 H23 N O4 . H Cl
Molekylvægt (g/mol) 365.85
Synonym Reticuline HCl
IUPAC navn (1S)-1-[(3-hydroxy-4-methoxyphenyl)methyl]-6-methoxy-2-methyl-3,4-dihydro-1H-isoquinolin-7-ol;hydrochloride
SMIL Cl.COc1ccc(C[C@@H]2N(C)CCc3cc(OC)c(O)cc23)cc1O
Tekniske data
Kemisk navn eller materiale Reticuline hydrochloride
Mængde 5 mg
InChI formel InChI=1S/C19H23NO4.ClH/c1-20-7-6-13-10-19(24-3)17(22)11-14(13)15(20)8-12-4-5-18(23-2)16(21)9-12;/h4-5,9-11,15,21-22H,6-8H2,1-3H3;1H/t15-;/m0./s1
Formel vægt 365.1394