Få mere at vide
Retapamulin, MedChemExpress
Artikelnummer. 16483277
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16483277 50 mg
16427268 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16483277 Leverandør Medchem Express Leverandørnr. HY17010
MedChemExpress Retapamulin(SB-275833) is a topical antibiotic, which binds to both E.
Retapamulin(SB-275833) is a topical antibiotic, which binds to both E.
Tekniske data
Kemisk navn eller materiale Retapamulin
Fysisk form Solid
CAS 224452-66-8
Farve White
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C30H47NO4S
Synonym SB-275833
Opløselighedsinformation DMSO : 110 mg/mL (212.45 mM; Need ultrasonic)
SMIL C[C@@H]1[C@]23[C@](C(CC3)=O)([H])[C@]([C@H](OC(CS[C@@H]4C[C@@H]5N(C)[C@@H](CC5)C4)=O)C[C@](C=C)(C)[C@H]1O)([C@H](C)CC2)C
Molekylvægt (g/mol) 517.76
Formel vægt 517.76
Procent renhed 98.0%