missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RET [V804M], GST Tag, His Tag Recombinant Protein
Human RET V804M, C-terminal fragment, amino acids H658-S1114 (as in NCBI/Protein entry NP_066124.1) with a V804M mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
3660.00 DKK - 55700.00 DKK
Specifications
Gene Alias | CDHF12, CDHR16, HSCR1, MEN2A, MEN2B, MTC1, PTC, RET51, RET-ELE1 |
---|---|
Protein Length | aa 658-1114 |
Species | Human |
Recombinant | Recombinant |
Protein Tag | GST Tag, His Tag |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478883
|
Invitrogen
PV6223 |
10μg |
3660.00 DKK
10µg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488883
|
Invitrogen
PV6224 |
100μg |
12470.00 DKK
100µg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15498883
|
Invitrogen
PV6225 |
1mg |
55700.00 DKK
1mg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
CDHF12, CDHR16, HSCR1, MEN2A, MEN2B, MTC1, PTC, RET51, RET-ELE1 | |
Human | |
GST Tag, His Tag |
aa 658-1114 | |
Recombinant |