missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RET Recombinant Human Protein
The receptor tyrosine kinase RET is a member of the cadherin superfamily.
Brand: Invitrogen™ PV4119
Additional Details : Weight : 0.00900kg
Description
RET plays a crucial role in neural crest development, and can undergo oncogenic activation by cytogenetic rearrangement.
Specifications
NP_066124 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
RET | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
79kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
RET Kinase Family | |
Recombinant | |
Baculovirus |