Få mere at vide
Resveratrol 3,4’-Diacetate, TRC
Artikelnummer. 30004405
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004405 5 mg
30003626 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004405 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1501305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 861446-14-2
Molekylær formel C18 H16 O5
Molekylvægt (g/mol) 312.317
Synonym 5-[(1E)-2-[4-(Acetyloxy)phenyl]ethenyl]-1,3-benzenediol 1-Acetate, 5-[(1E)-2-[4-(Acetyloxy)phenyl]ethenyl]-1,3-benzenediol Monoacetate
IUPAC navn [4-[(E)-2-(3-acetyloxy-5-hydroxyphenyl)ethenyl]phenyl] acetate
SMIL CC(=O)Oc1ccc(\C=C\c2cc(O)cc(OC(=O)C)c2)cc1
Tekniske data
Kemisk navn eller materiale Resveratrol 3,4'-Diacetate
Mængde 5 mg
InChI formel InChI=1S/C18H16O5/c1-12(19)22-17-7-5-14(6-8-17)3-4-15-9-16(21)11-18(10-15)23-13(2)20/h3-11,21H,1-2H3/b4-3+
Formel vægt 312.1