missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Moldex™ Smart Solo™ FFP2 NR D Respirator Mask with Ventex™-Valve and Special Strap System
Click to view available options
:
Smart Solo™ FFP2 NR D Respirator Mask with Ventex™-Valve and Special Strap System
Description
- FFP2 NR D Protection
- Single use respirator face mask with valve
- Fitted with Ventex™-valve
- Reliable protection against particles
- Specially designed head-piece - one retaining strap
- ActivForm™ 3D design adapts to face shape ensuring comfortable and secure fit
- DuraMesh™ structure preserve the mask shape under testing conditions
Specifications
Specifications
Certifications/Compliance (Europe) | FFP2 NR D, EN 149:2001+A1:2009, (EU)2016/425 |
Design Type | Cup/Molded |
Material | Polypropylene, EVA |
With Exhalation Valve | Yes |
Filtration Layer Material | Polypropylene |
For Use With (Equipment) | Hazardous fine dusts, water and oil based mists, aerosols, biological agents of risk group 2 |
Material Headstrap | Polyester, Lycra |
Nose Piece Material | Polyethylene |
Nosepiece Type | Flexible nosepiece |
Strap Attachment Type | 1-Strap with adjustable head harness |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Moldex™ Smart Solo™ FFP2 NR D Respirator Mask with Ventex™-Valve and Special Strap System >
Spot an opportunity for improvement?Share a Content Correction