Få mere at vide
Resorufin Benzyl Ether, TRC
Artikelnummer. 30416213
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416213 10 mg
30428741 5 mg
30415591 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30416213 Leverandør Toronto Research Chemicals Leverandørnr. TRCR14651810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 87687-02-3
Molekylær formel C19H13NO3
Molekylvægt (g/mol) 303.31
Synonym 7-(Phenylmethoxy)-3H-phenoxazin-3-one, 7-Benzyloxyphenoxazone, 7-Benzyloxyresorufin
IUPAC navn 7-(benzyloxy)-3H-phenoxazin-3-one
SMIL O=C1C=CC2=NC3=C(OC2=C1)C=C(OCC4=CC=CC=C4)C=C3
Tekniske data
Kemisk navn eller materiale Resorufin Benzyl Ether
Mængde 10 mg
InChI formel InChI=1S/C19H13NO3/c21-14-6-8-16-18(10-14)23-19-11-15(7-9-17(19)20-16)22-12-13-4-2-1-3-5-13/h1-11H,12H2
Formel vægt 303.09