Learn More
Resorcinol, TRC
Product Code. 30041307
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041307 1 g 1g
30003827 100 g 100g
30007081 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30041307 Supplier Toronto Research Chemicals Supplier No. TRCR1447001G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-46-3
Molecular Formula C6 H6 O2
Molecular Weight (g/mol) 110.11
Synonym Resorcinol, 1,3-Dihydroxybenzene, Fluorescein Sodium Imp. A (EP), Hymecromone Imp. A (EP), Hexylresorcinol Imp. B (EP), 1,3-Benzenediol, Resorcinol (8CI), 1,3-Phenylenediol, 3-Hydroxyphenol, C.I. 76505, C.I. Developer 4, C.I. Oxidation Base 31, Developer O, Developer R, Developer RS, Durafur Developer G, Fouramine RS, Fourrine 79, Fourrine EW, NSC 1571, Nako TGG, Pelagol Grey RS, Pelagol RS, RS 11H, RS 11L, Redimix 401RAP60, Reso, Resorcin, Resorcinol 80, Rezorsine, Rodol RS, m-Benzenediol, m-Dihydroxybenzene, m-Hydroquinone, m-Hydroxyphenol, m-Hydroxyphenol, m-Phenylenediol, m-Resorcinol
IUPAC Name benzene-1,3-diol
SMILES Oc1cccc(O)c1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H
Formula Weight 110.0368
Percent Purity >95%
Chemical Name or Material Resorcinol (1,3-Benzenediol)
Grade HPLC