Få mere at vide
Resorantel, LoGiCal
Artikelnummer. 30463886
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463886 10 mg
1 options
Artikelnummer. 30463886 Leverandør LoGiCal Leverandørnr. LGCFOR3223.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Resorantel
CAS 20788-07-2
Analyt- eller komponentnavne Resorantel
Forsendelsestilstand Room Temperature
Molekylær formel C13 H10 Br N O3
Certificeringer/compliance ISO 9001
Synonym Benzamide, N-(4-bromophenyl)-2,6-dihydroxy-, 2,6-Dihydroxybenzoic acid 4'-bromanilide, 4'-Bromo-2,6-dihydroxybenzanilide, HOE 296V, Resorantel, Resorcylam, Terenol, gamma-Resorcylanilide, 4'-bromo- (8CI), 4'-Bromo-gamma-resorcylanilide
InChI formel InChI=1S/C13H10BrNO3/c14-8-4-6-9(7-5-8)15-13(18)12-10(16)2-1-3-11(12)17/h1-7,16-17H,(H,15,18)
Løsningstype Neat
SMIL Oc1cccc(O)c1C(=O)Nc2ccc(Br)cc2
IUPAC navn N-(4-bromophenyl)-2,6-dihydroxybenzamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 308.13
Mængde 10 mg
Formel vægt 306.9844 g/mol