Få mere at vide
Resiniferatoxin, 99+%, Thermo Scientific™
Artikelnummer. p-7042415
Skift visning
Mængde:
1 mg
10 mg
Pakningsstørrelse:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
15404839 1 mg 1mg
15414839 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 15404839 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. J62458.M^
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Resiniferatoxin, a potent capsaicin-like stimulator of peripheral nociceptors in the neonatal rat tail in vitro. Resiniferatoxin is used as an anti-inflammatory, anti-viral and anti-psoriatic uses. Diterpene ester related to phorbol but apparently not tumorigenic activates protein kinase C
Solubility
Soluble in DMSO to 100mM and to 50mM in ethanol
Notes
Resiniferatoxin (RTX) is susceptible to slow degradation if exposed to air. Even at -20°C, Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep in a dry place. Keep away from strong oxidizing agents. RTX must be stored in an inert atmosphere for long-term stability.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57444-62-9
Molekylær formel C37H40O9
Molekylvægt (g/mol) 628.72
MDL nummer MFCD00135927,MFCD00135927
InChI nøgle OYWHJFQVSOUQSF-PKRQWDGKSA-N
Synonym resiniferatoxin
PubChem CID 57090162
IUPAC navn [(1R,2R,6R,10S,11R,15R,17R)-13-benzyl-6-hydroxy-4,17-dimethyl-5-oxo-15-(prop-1-en-2-yl)-12,14,18-trioxapentacyclo[11.4.1.0¹,¹⁰.0²,⁶.0¹¹,¹⁵]octadeca-3,8-dien-8-yl]methyl 2-(4-hydroxy-2-methoxyphenyl)acetate
SMIL COC1=CC(O)=CC=C1CC(=O)OCC1=C[C@H]2[C@H]3OC4(CC5=CC=CC=C5)O[C@]3(C[C@@H](C)[C@]2(O4)[C@@H]2C=C(C)C(=O)[C@@]2(O)C1)C(C)=C
Tekniske data
Kemisk navn eller materiale Resiniferatoxin
Farve White
Mængde 1 mg
FN nummer UN2811
Beilstein 5371150
Opløselighedsinformation Soluble in DMSO to 100mM and to 50mM in ethanol
Formel vægt 628.71
Procent renhed ≥99%
Fysisk form Solid