Få mere at vide
Reserpine-d9, TRC
Artikelnummer. 30409370
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30409370 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409370 Leverandør Toronto Research Chemicals Leverandørnr. TRCR14460225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Reserpine trimethoxy-d9
CAS 84759-11-5
Renhedsgrad noter HPLC
Molekylær formel C33 2H9 H31 N2 O9
Mængde 25 mg
Synonym Yohimban-16-carboxylic acid','11,17-dimethoxy-18-[[3,4,5-tri(methoxy-d3)benzoyl]oxy]-','methyl ester','(3β,16β,17α,18β,20α)- (9CI), Benz[g]indolo[2,3-a]quinolizine','yohimban-16-carboxylic acid deriv.
InChI formel InChI=1S/C33H40N2O9/c1-38-19-7-8-20-21-9-10-35-16-18-13-27(44-32(36)17-11-25(39-2)30(41-4)26(12-17)40-3)31(42-5)28(33(37)43-6)22(18)15-24(35)29(21)34-23(20)14-19/h7-8,11-12,14,18,22,24,27-28,31,34H,9-10,13,15-16H2,1-6H3/t18-,22+,24-,27-,28+,31+/m1/s1/i2D3,3D3,4D3
SMIL [2H]C([2H])([2H])Oc1cc(cc(OC([2H])([2H])[2H])c1OC([2H])([2H])[2H])C(=O)O[C@@H]2C[C@@H]3CN4CCc5c([nH]c6cc(OC)ccc56)[C@H]4C[C@@H]3[C@@H]([C@H]2OC)C(=O)OC
IUPAC navn methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-[3,4,5-tris(trideuteriomethoxy)benzoyl]oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
Molekylvægt (g/mol) 617.73
Formel vægt 617.3299
Procent renhed >95