Få mere at vide
Eppendorf™ Research™ plus BASIC - Trade-In Pipette
GREENER_CHOICE
Artikelnummer. p-200132151 Shop alle Eppendorf produkter
Volumen (metrisk):
0.1 to 2.5 μL
0.25 to 2.5 mL
0.5 to 10 μL
0.5 to 5 mL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Farve:
Blue
Dark Gray
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Pakningsstørrelse:
Hver
Artikelnummer. 15733449
Brand: Eppendorf™ 3123000217
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Adapt for specific liquids or other external conditions with ultra-light fully autoclavable pipettes
  • Volume adjustment feature allows desired volume to be reached in few quick turns
  • Control button and ejectors low operating force are positioned for ergonomics 4-digit, magnifying-shape volume display
  • Calibration display and adjustment opening for specific liquids and volumes
  • Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
  • Quick lock makes lower part particularly easy to remove and service
  • Spring loaded tip cone promotes tight fit to tip
  • Has ability to be autoclaved without disassembling
  • Applications: Pipetting liquids, Mixing liquids, Filling of plates, gels, and reaction vessels, Phase extraction and removal of supernatants
TRUSTED_SUSTAINABILITY
Tekniske data
Certificeringer/compliance Lot Specific Certificate
Kompatible tips epT.I.P.S.
Justerbar spidsafstand No
Autoklaverbar Autoclavable
Farve Dark Gray
Engangs Reusable
Til brug med epT.I.P.S. 10 μL
Greb Type Ergonomic
Materiale Plastic
Antal kanaler 1
Programmerbarhed No
Sterilitet Non-sterile
Type Manual Pipette
Volumen (metrisk) 0.1 to 2.5 μL
Til brug med (applikation) Pipette Tips
Til brug med (udstyr) epT.I.P.S.Pipette Tips
Inkluderer epT.I.P.S.Pipette Tips