missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies RepSox
Beskrivelse
- RepSox is a cell-permeable, selective inhibitor of the TGF-β type 1 receptor (TGFβRI) ALK5 (IC50 = 4, 18, and 23 nM for ALK5 autophosphorylation, TGF-β cellular assay, and ALK5 binding in HepG2 cells, respectively; Gellibert et al.) This inhibitor demonstrates less potent activity (IC50 > 16 μM) against 9 related kinases, including p38 MAPK and GSK3 (Gellibert et al.)
- Enhances reprogramming of mouse embryonic fibroblasts (MEFs) that have been transduced with OCT4, KLF4, and c-MYC (Ichida et al.; Subramanyam et al.)
- Direct lineage reprogramming of fibroblasts to mature neurons, in combination with CHIR99021, VaLproic Acid, Forskolin, SP600125, Gö6983 and Y-27632 (Hu et al.) DIFFERENTIATION
- Alone or in combination with Forskolin, Dexamethasone, and Nicotinamide, induces differentiation of human pancreatic progenitor cells into insulin-producing cells (Kunisada et al.; Rezania et al.)
Tekniske data
Tekniske data
| CAS | 446859-33-2 |
| Synonym | ALK5 inhibitor II; E-616452; SJN 2511 |
| Mængde | 1 mg |
| Molekylær formel | C17H13N5 |
| Renhed | ≥98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?