Få mere at vide
Reproterol Hydrochloride, LoGiCal
Beskrivelse
Reproterol Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Reproterol Hydrochloride |
| CAS | 13055-82-8 |
| Analyt- eller komponentnavne | Reproterol Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H23 N5 O5 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1 H-Purine-2,6-dione, 7-[3-[[2-(3,5-dihydroxyphenyl)-2-hydroxyethyl]amino]propyl]-3,7-dihydro-1,3-dimethyl-, hydrochloride (1:1), 1 H-Purine-2,6-dione, 7-[3-[[2-(3,5-dihydroxyphenyl)-2-hydroxyethyl]amino]propyl]-3,7-dihydro-1,3-dimethyl-, monohydrochloride (9 CI), Asmaterolo, Bronchodil, Bronchospasmin, Theophylline, 7-[3-[(beta,3,5-trihydroxyphenethyl)amino]propyl]-, monohydrochloride (8 CI), Theophylline, 7-[3-[(beta,3,5-trihydroxyphenethyl)amino]propyl]-, monohydrochloride (8 CI), Theophylline, 7-[3-[(beta,3,5-trihydroxyphenethyl)amino]propyl]-, monohydrochloride (8 CI), Theophylline, 7-[3-[(beta,3,5-trihydroxyphenethyl)amino]propyl]-, monohydrochloride (8 CI), Theophylline, 7-[3-[(beta,3,5-trihydroxyphenethyl)amino]propyl]-, monohydrochloride (8 CI) |
| InChI formel | InChI=1 S/C18H23N5O5.ClH/c1-21-16-15(17(27)22(2)18(21)28)23(10-20-16)5-3-4-19-9-14(26)11-6-12(24)8-13(25)7-11;/h6-8,10,14,19,24-26 H,3-5,9H2,1-2H3;1 H |
| SMIL | Cl.CN1C(=O)N(C)c2ncn(CCCNCC(O)c3cc(O)cc(O)c3)c2C1=O |
| IUPAC navn | 7-[3-[[2-(3,5-dihydroxyphenyl)-2-hydroxyethyl]amino]propyl]-1,3-dimethylpurine-2,6-dione;hydrochloride |
| Vis mere |