Få mere at vide
Reposal, LoGiCal
Artikelnummer. 30461338
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461338 10 mg
1 options
Artikelnummer. 30461338 Leverandør LoGiCal Leverandørnr. LGCFOR1615.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Reposal
CAS 3625-25-0
Analyt- eller komponentnavne Reposal
Forsendelsestilstand Room Temperature
Molekylær formel C14 H18 N2 O3
Certificeringer/compliance ISO 9001
Synonym 2,4,6(1H,3H,5H)-Pyrimidinetrione, 5-bicyclo[3.2.1]oct-2-en-3-yl-5-ethyl-Barbituric acid, 5-bicyclo[3.2.1]oct-2-en-3-yl-5-ethyl- (7CI,8CI), 5-Bicyclo[3,2,1]oct-2-en-2-yl-5-ethylbarbituric acid, 5-Bicyclo[3.2.1]oct-2-en-3-yl-5-ethylbarbituric acid, Reposal, Reposal Novo, Reposamal, WT 161
InChI formel InChI=1S/C14H18N2O3/c1-2-14(11(17)15-13(19)16-12(14)18)10-6-8-3-4-9(5-8)7-10/h6,8-9H,2-5,7H2,1H3,(H2,15,16,17,18,19)
Løsningstype Neat
SMIL CCC1(C(=O)NC(=O)NC1=O)C2=CC3CCC(C3)C2
IUPAC navn 5-(3-bicyclo[3.2.1]oct-3-enyl)-5-ethyl-1,3-diazinane-2,4,6-trione
Industry Type Clinical Testing
Molekylvægt (g/mol) 262.3
Mængde 10 mg
Formel vægt 262.1317 g/mol