Learn More
Oakton™ Replacement Reagents for Colorimeters
High quality powder reagents for exceptional accuracy, repeatablilty, and stability.
Brand:  Oakton™ 35645-64
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 15634839
  • 685.00 DKK / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
Applications: Electrochemical, Water Quality, Life Sciences
Specifications
Specifications
EPA, DPD
50
Free Chlorine Reagent
C201, C301, C401 Oakton™ colorimeters
0.01 to 1.99ppm