Få mere at vide
Labconco™ Udskiftning af O-ring
Artikelnummer. 16224971 Shop alle Labconco Corporation produkter
Skift visning
For Use With: Precise Glove Boxes, Protector Combination Glove Boxes, XPert Weigh Boxes
Artikelnummer.
16224971 For Use With: Precise Glove Boxes, Protector Combination Glove Boxes, XPert Weigh Boxes
1 options
Artikelnummer. 16224971 Leverandør Labconco™ Leverandørnr. 1640000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Replacement O-Ring is for use with various glove boxes.
Applicable Products:
  • Precise Basic Glove Boxes
  • Precise Controlled Atmosphere Glove Boxes
  • Precise HEPA-Filtered Glove Boxes
  • Protector Combination Glove Boxes
  • Protector Controlled Atmosphere Glove Boxes
  • Protector Filtered Glove Boxes
  • XPert Weigh Boxes
Tekniske data
Til brug med (udstyr) Precise Basic Glove Boxes, Precise Controlled Atmosphere Glove Boxes, Precise HEPA-Filtered Glove Boxes, Protector Combination Glove Boxes, Protector Controlled Atmosphere Glove Boxes, Protector Filtered Glove Boxes, XPert Weigh Boxes
Vægt (engelsk) 1 lb.
Vægt (metrisk) 0.5 Kg
Produkttype Replacement O-Ring